ContactUsLinkComponent
TDS Standard, Environmental Express
TDS Standard, Environmental Express
Catalog # 76436-958
CAS Number:  
undefined
TDS Standard, Environmental Express
Catalog # 76436-958
Supplier Number:  F95010
CAS Number:  
Specifications
  • Description:
    TDS Standard
  • Size:
    10 pk
  • Cat. No.:
    76436-958
Specifications
About this item
Save time and money with a TDS standard compliant to SM 2540 and ASTM D5907 methods.
The Total Dissolved Solids (TDS) standard provides ongoing batch level QA/QC samples for the TDS analysis at an affordable cost. Create a Lab Control Sample (LCS) by transferring bottle contents into a graduated cylinder or volumetric flask, pour to volume and analyze. An acceptability range of ±3 standard deviations is provided for each parameter.
Each bottle of standard has a unique ID# and corresponding unique solids values.
Delivery: MSDS and Certificate of Analysis with NIST traceability and instructions for use are included with each set.