Order Entry
Puerto Rico
ContactUsLinkComponent
 
Scandium Single-Element Organometallic Oil Standard for AA and ICP, SPEX CertiPrep
  SXORG-SC8-2Z
 :  
Scandium Single-Element Organometallic Oil Standard for AA and ICP, SPEX CertiPrep
  SXORG-SC8-2Z
 :  ORG-SC8-2Z
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    Base Oil 75
  • Pack type:
    LDPE bottle
  • Size:
    50 g
  • Cat. No.:
    SXORG-SC8-2Z
  • Element:
    Scandium

 

 

Organometallic single-element oil standards are guaranteed stable and accurate, together with a wide range of applications and features:

  • Wear metals
  • Crude oils
  • Additive metals
  • Environmental monitoring Petrochemical testing
  • Pharmaceuticals
  • Food processing
  • Sulfur in diesel fuel

The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

 : Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025. Certificate of analysos with every solution