Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium (K+) Single-Element Ion Cation Standard, SPEX CertiPrep
Potassium (K+) Single-Element Ion Cation Standard, SPEX CertiPrep
  SXCS-K2-2Y
 :  
Potassium (K+) Single-Element Ion Cation Standard, SPEX CertiPrep
  SXCS-K2-2Y
 :  CS-K2-2Y
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    0.2% HNO₃
  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Cat. No.:
    SXCS-K2-2Y
  • Element:
    Potassium

 

 

Single-element standards for ion chromatography and ion selective electodes.

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.