Order Entry
United States
ContactUsLinkComponent
Claritas PPT® Grade Minerals Testing Kit, SPEX CertiPrep
Claritas PPT® Grade Minerals Testing Kit, SPEX CertiPrep
Catalog # SXSPXMT-KIT
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade Minerals Testing Kit, SPEX CertiPrep
Catalog # SXSPXMT-KIT
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  SPXMT-KIT
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    LDPE bottle
  • Description:
    Minerals Testing Kit,
  • Size:
    30 ml
  • Environmentally Preferable:
  • Cat. No.:
    SXSPXMT-KIT

Specifications

About this item

Claritas PPT® Grade Minerals Testing Kit for ICP-MS are designed for routinely analyzed minerals.

Kits come with six, 30 mL standards which include a nitric acid blank for easy dilution. Conveniently packaged in a sturdy, heavy-duty carton, these kits are perfect to store on a lab bench or in a cabinet.

The 30 mL standards ship non-hazardous, saving money on shipping costs. The smaller volume also allows for less hazardous waste, should the standard expire before its contents are used.