Order Entry
Puerto Rico
ContactUsLinkComponent
Instrument Calibration Standard 3, SPEX CertiPrep
Instrument Calibration Standard 3, SPEX CertiPrep
Catalog # SXICAL-3
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Instrument Calibration Standard 3, SPEX CertiPrep
Catalog # SXICAL-3
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  ICAL-3
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    LDPE bottle
  • Description:
    Instrument Calibration Standard 3
  • Size:
    125 ml
  • Cat. No.:
    SXICAL-3

Specifications

About this item

CLP Standards for ICP & ICP-MS

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

Instrument Calibration for CLP Methods
May be used separately or mixed together for preparation of the analytical curve. When mixed, these solutions will yield a standard containing all of the elements in the Target Analyte List (TAL). Instruments must be calibrated daily, every 24 hours, or each time the instrument is set-up. Calibration standards must be prepared fresh for each analysis and discarded after use. A dilution of 100-fold is suggested. Antimony and mercury can be diluted as required.

For ISM 01.2, at least one of your calibration standards must be at the Contract Required Quantification Limit (CRQL). See ISM 01.2 sections for CRQL standards