Order Entry
United States
ContactUsLinkComponent
 
Initial Calibration Verification Standard 1A, SPEX CertiPrep
  SXICV-1A
 :  
Initial Calibration Verification Standard 1A, SPEX CertiPrep
  SXICV-1A
 :  ICV-1A
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Matrix:
    5% HNO₃/tr. Tartaric acid/tr. HF
  • Pack type:
    LDPE bottle
  • Description:
    Initial Calibration Verification Standard 1A
  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    SXICV-1A

 

 

CLP Standards for ICP & ICP-MS
Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

Initial Calibration Verification for CLP Methods
The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods. The CLP methods require calibration of analytical instrumentation whithin the expected quantitative range (ICAL standards) and additional CLP QA standards (ICV standards) to verify the calibration curve at each of the selected wavelengths that wil be used for sample analysis.

Our verification standards contain all of the elements on the TAL list and are independent standards for testing an instruments calibration curve. SPEX CertiPrep's ICV standards are designed to be used with their corresponding instrument calibration standards (ICAL).

We recommend dilution of ICV standards to a range within your instruments calibration curve. A dilution of 20-fold is recommended.