Order Entry
Puerto Rico
ContactUsLinkComponent
ICP Spike Sample 4, SPEX CertiPrep
ICP Spike Sample 4, SPEX CertiPrep
Catalog # SXSPIKE-4
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
ICP Spike Sample 4, SPEX CertiPrep
Catalog # SXSPIKE-4
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  SPIKE-4
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr. Tartaric acid/tr. HF
  • Pack type:
    LDPE bottle
  • Description:
    ICP Spike Sample 4
  • Size:
    125 ml
  • Cat. No.:
    SXSPIKE-4

Specifications

About this item

CLP Standards for ICP & ICP-MS
Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

CLP ISM 01.2 and ILM 05.2 & 05.3 Standards for ICP
Our spike standard, SPIKE-4, provides all of the analytes required for the IC, ICP-AES and the AA spike.