To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml
- Matrix:Multiple
- Pack type:LDPE bottle
- Description:Heavy Metals Testing Kit,
- Size:30 ml
- Environmentally Preferable:
- Cat. No.:SXSPXHM-KIT
Specifications
About this item
Heavy Metals & Minerals Testing Kits
For routinely analyzed Heavy Metals and Minerals
Heavy Metals and Minerals Testing Kits are designed for routinely analyzed heavy metals and minerals. All kits come with six, 30 ml standards which includes a nitiric acid blank for easy dilution. Conveniently packaged in a sturdy, heavy-duty carton, these kits are perfect to store on a lab bench or in a cabinet. The 30 ml standards ship non-hazardous, saving money on shipping costs. The smaller volume also allows for less hazardous waste should the standard expire before its contents are used.