Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Elemental Impurities E, SPEX CertiPrep

Elemental Impurities E, SPEX CertiPrep

Elemental Impurities E, 125 ml, 5% HNO₃, LDPE bottle
Catalog # SXICH-TXM7
Supplier:   SPEX CERTIPREP LLC
Supplier #:  ICH-TXM7
Elemental Impurities E, SPEX CertiPrep
Catalog # SXICH-TXM7
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Elemental Impurities E, SPEX CertiPrep

Elemental Impurities E, 125 ml, 5% HNO₃, LDPE bottle

CAS Number:  
Price
$319.56
Log in for availability
Each (125ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

ICH Standard

The new guidelines set by the Internatioal Conference on Harmonization (ICH) have pushed the pharmaceutical and nutraceutical industries to provide accurate, quantifiable results for metal analysis in drugs, pharmaceutical substances and raw materials.

USP <232> outlines new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury. The procedures forcus on the use of ICP-MS for the analysis of low level impurities. ICP-MS instrumentation, along with accurate ICP-MS standards, allow for increased efficiency and accuracy of the analysis necessary to comply with the new regulations. In addition to the changes enacted by the USP.

Developed in accordance with USP <232> Elemental Impurities, SPEX CertiPrep offers hese additions to our Consumer Safety Compliance Standards line. These standards can be used as a calibration or check standard to verify Oral Daily Dose PDE, Parenteral Component Limit or Parenteral Daily Dose PDE as well as Inhalation Component Limit or Daily Dose. Our extensive experience in creating quality trace metal standards, coupled with your ICP-MS analysis, will ensure your company will remain compliant with the new and changing regulations.

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    LDPE bottle
  • Description:
    Elemental Impurities E
  • Size:
    125 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.