To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Matrix:2% HNO₃
- Pack type:LDPE bottle
- Description:ICP-MS Interferents Check Solution B2 (for SW-846)
- Size:125 ml
- Cat. No.:SXCL-INT-B2
Specifications
About this item
CLP interferents standards for ICP & ICP-MS
Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.
The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.
US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
- Exhibit C, Inorganic Target Analyte List (TAL)
- Exhibit D, Analytical Methods
- Exhibit E, QA/QC Requirements
Alternate Standards
We also provide a solution of alternate interferents and alternate analysis. Alternate interferents A (INT-A2) and alternate analytes B (INT-B2) may be prepared in combination with the INT-A1 and INT-B3 solutions mentioned, or any combination involving the four solutions, depending on the analytes and interferents of interest to you. We provides ICP-MS interferents and interferent check solutions for SW-845.