Claritas PPT® Grade Zinc 68 Standards for ICP-MS, SPEX CertiPrep
Catalog # SXISOT-ZN68
Supplier: SPEX CERTIPREP LLC
CAS Number:
Specifications
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Pack type:LDPE bottle
- Size:125 ml
- Cat. No.:SXISOT-ZN68
- Element:Zinc 68
Specifications
About this item
Single element standards for ICP-MS.
- Made with an acid and ASTM Type I Water
- Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
- Directly traceable to NIST (where applicable)
Claritas PPT® Grade CRMs are designed for ICP and ICP-MS analysis. They are available in single and multi-element solutions. The standards are set at 1 µg/ml, 100 µg/ml, or 1,000 µg/ml and packaged in 30 ml and 125 ml bottles to minimize contamination. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.
Certifications: Certified to DQS to ISO 9001:2015
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016