Order Entry
Puerto Rico
ContactUsLinkComponent
 
Claritas PPT® Grade ICP-MS Spike Sample Standard 2 (Soil), SPEX CertiPrep
Catalog # SXCL-SPIKE-2
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Claritas PPT® Grade ICP-MS Spike Sample Standard 2 (Soil), SPEX CertiPrep
Catalog # SXCL-SPIKE-2
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CL-SPIKE-2
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Matrix:
    5% HNO₃/tr. Tartaric acid/tr. HF
  • Pack type:
    LDPE bottle
  • Description:
    ICP-MS Spike Sample Standard 2 (Soil)
  • Size:
    125 ml
  • Cat. No.:
    SXCL-SPIKE-2

Specifications

About this item

as PPT® multi-element spike standards for ICP-MS

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.

Spike Sample Analysis: Designed for addition to a matrix blank prior to digestion for both water and soil. An aliquot of the respective Spike Standard should be added to produce the proper concentration levels in the digestate.

CLP-M/6020/SW-846 Standards for ICP-MS. The spike standards, CL-SPIKE-1 and CL-SPIKE-2, provide all of the analytes required for ICP-MS. Add 1 mL of CL-SPIKE-1 to aqueous samples and 2 mL of CL-SPIKE-2 to solid samples prior to digestion.

Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.