Order Entry
United States
ContactUsLinkComponent
 
Claritas PPT® Grade ICP-MS Multi-Element Solution Standards Set with Mercury, SPEX CertiPrep
  SXCLMS-SET
 :  
Claritas PPT® Grade ICP-MS Multi-Element Solution Standards Set with Mercury, SPEX CertiPrep
  SXCLMS-SET
 :  CLMS-SET
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Concentration:
    10 µg/ml
  • Matrix:
    Multiple
  • Pack type:
    LDPE bottle
  • Description:
    ICP-MS Multi-Element Solution Standards Set with Mercury
  • Size:
    125 ml
  • Environmentally Preferable:
  • Cat. No.:
    SXCLMS-SET

 

 

Claritas PPT® multi-element standards for ICP-MS.

SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.

The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.

Multi-Solution Solution Standards: Designed to contain virtually every element in the mass spectrum for concentration verification checks.

 : SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.