Order Entry
Puerto Rico
ContactUsLinkComponent
Assurance® Grade Groundwater and Wastewater Pollution Control Check Standards, Set of 3 Trace Metals Standards with Mercury for AA & ICP, SPEX CertiPrep
Assurance® Grade Groundwater and Wastewater Pollution Control Check Standards, Set of 3 Trace Metals Standards with Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXTM-SET
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Assurance® Grade Groundwater and Wastewater Pollution Control Check Standards, Set of 3 Trace Metals Standards with Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXTM-SET
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  TM-SET
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Matrix:
    HNO₃ dil.
  • Pack type:
    LDPE bottle
  • Description:
    Groundwater and Wastewater Pollution Control Check Standards, Set of 3 Trace Metals Standards with Mercury
  • Size:
    3x 125 ml
  • Cat. No.:
    SXTM-SET

Specifications

About this item

Groundwater and wastewater pollution control check standards. Assurance® mixed multi-element standards for AA & ICP.

Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.

May be used either as standards or as a means to check the individual analysts accuracy and precision.

Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods.