ContactUsLinkComponent
Assurance® Grade Interference Check Multi-Element Standards without Mercury for AA & ICP, SPEX CertiPrep
Assurance® Grade Interference Check Multi-Element Standards without Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXINTER-SETN
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Assurance® Grade Interference Check Multi-Element Standards without Mercury for AA & ICP, SPEX CertiPrep
Catalog # SXINTER-SETN
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  INTER-SETN
CAS Number:  
Specifications
  • Pack type:
    LDPE bottle
  • Description:
    Interference Check Multi-Element Standards without Mercury
  • Size:
    3x 125 ml
  • Cat. No.:
    SXINTER-SETN
Specifications
About this item
Assurance® mixed multi-element interference check standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
The Interference Check Standards are used to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).