Order Entry
United States
ContactUsLinkComponent
Anti-DsRed Rabbit Polyclonal Antibody
Anti-DsRed Rabbit Polyclonal Antibody
Catalog # 76309-344
undefined
Anti-DsRed Rabbit Polyclonal Antibody
Catalog # 76309-344
Supplier Number:  600-401-379-RTU
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    RFP
  • Antigen Symbol:
    DsRed
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Form:
    Liquid (sterile filtered)
  • Antigen Synonyms:
    DsRed protein, rDsRed, Discosoma sp. Red Fluorescent Protein, Red fluorescent protein drFP583, sea anemone Discosoma sp. Mushroom, RFP antibody, RFP-RTU, Ready To Use Antibody, RTU Antibody
  • Storage Buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Shipping Temperature:
    Wet Ice
  • Purification:
    Affinity Purified
  • Cat. No.:
    76309-344

Specifications

About this item

Fluorescent proteins such as green fluorescent protein (GFP) or Discosoma Red Fluorescent Protein (or DsRFP) are widely used in research practice. DsRed is isolated from sea anemone Discosoma sp. mushroom. GFP and DsRed share only 19% identity, therefore, in general, anti-GFP antibodies do not recognize DsRed protein and vice versa. Structurally, DsRed protein is similar to GFP protein in terms of its overall fold (a β-can) and chromophore-formation chemistry. However, GFP protein undergoes an additional step in the chromophore maturation and obligates tetrameric structure. Using site-directed mutagenesis, several DsRed protein variants have been created allowing the red fluorescent protein mature as a monomeric form. Among DsRFP monomeric variants are monomeric mutant mRFP1, mBanana, mCherry, mHoneydew, mPlum, mOrange, mStrawberry and mTangerine that offer a wide range of fluorescent colors. As Rockland RFP polyclonal antibodies are raised against whole RFP protein of wild type, the polyclonal antibodies are expected to recognize all RFP variant forms. This RFP antibody has been pre-absorbed to eliminate any potential cross-reactivity to human, mouse and rat serum proteins. The antibodies are also confirmed for non-reactivity to GFP protein. All Rockland Immunochemical’s RFP antibodies are affinity-purified to assure both high affinity and specificity. Rigorous quality control testing ensures that the finished product meets or exceeds out high standards for optimum performance in your assays.

Type: Primary
Antigen: DsRed
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: