Mouse Recombinant VEGF164 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized VEGFR1/R2, Fc Tag at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.02-0.5 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human VEGF R1, His Tag (Cat. No. VE1-H5220) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.2-2 ng/mL (Routinely tested).
- Gene ID:Q00731-2
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:RP1-261G23.1|MVCD1|MGC70609|VEGFA|VPF
- Protein/Peptide Name:VEGF164
- Purity:>95%
- Molecular Weight:22.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together