Mouse Recombinant PLGF (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 100 mM HAC, pH2.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-M5256) at 2 μg/mL (100 μL/well) can bind Mouse PLGF, His Tag (Cat. No. PGF-M52H0) with a linear range of 0.2-4 ng/mL (QC tested).
- Gene ID:P49764-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:PGF|PLGF|PlGF2|PGFL|PlGF|SHGC-10760
- Protein/Peptide Name:PLGF
- Purity:>95%
- Molecular Weight:17.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 100 mM HAC, pH2.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together