Mouse Recombinant Neuropilin-1 (from HEK293)
Catalog # 103739-282
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse NRP1, Mouse IgG2a Fc Tag (Cat. No. NR1-M5254) with a linear range of 0.5-4 ng/mL (QC tested).
- Gene ID:P97333-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:NRP1|Neuropilin-1|VEGF165R|NRP|CD304
- Protein/Peptide Name:Neuropilin-1
- Purity:>90%
- Molecular Weight:120.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-282