Mouse Recombinant LAIR-1 (from HEK293)
Catalog # 103738-256
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LA1-M5254) with a linear range of 1-10 ng/mL (QC tested).
- Gene ID:Q8BG84-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:LAIR1|CD305
- Protein/Peptide Name:LAIR-1
- Purity:>95%
- Molecular Weight:40.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-256