Mouse Recombinant FOLR1 (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Folic Acid-BSA Conjugate at 1 μg/mL (100 μL/well) can bind Mouse FOLR1, His Tag (Cat. No. FO1-M5225) with a linear range of 0.02-0.313 μg/mL (QC tested).
- Gene ID:P35846-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FOLR-1|FOLR|FBP
- Protein/Peptide Name:FOLR1
- Purity:>95%
- Molecular Weight:26.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together