Mouse Recombinant CD38 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM MES, 100 mM NaCl, pH6.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P56528-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD38|T10|cADPr hydrolase 1
- Protein/Peptide Name:CD38
- Purity:>90%
- Molecular Weight:31.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM MES, 100 mM NaCl, pH6.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together