Mouse Recombinant CD155 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Mouse TIGIT, Fc Tag (Cat. No. TIT-M5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse CD155, Fc,Avitag (Cat. No. CD5-M82F7) with a linear range of 20-313 ng/mL (QC tested).
- Gene ID:Q8K094-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/Peptide Name:CD155
- Purity:>95%
- Molecular Weight:63.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together