Human Recombinant TRAIL R2 (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) at 10 μg/mL (100 μL/well) can bind Biotinylated Human TRAIL R2, Avitag,His Tag (Cat. No. TR2-H82E6) with a linear range of 2-78 ng/mL (QC tested).
- Gene ID:O14763-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TRAILR2|TRAIL-R2|CD262|TRICK2|KILLER|TRICKB|ZTNFR9|TNFRSF10B|DR5
- Protein/Peptide Name:TRAIL R2
- Purity:>90%
- Molecular Weight:16.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT