Human Recombinant TNFSF11 (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Captured Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) on CM5 chip via Anti-Mouse antibodies surface can bind Human TNFSF11, Fc Tag (Cat. No. RAL-H5265) with an affinity constant of 0.574 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) on AMC Biosensor, can bind Human TNFSF11, Fc Tag (Cat. No. RAL-H5265) with an affinity constant of 0.205 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:OPGL|ODF|TRANCE|CD254|RANKL
- Protein/Peptide Name:TNFSF11
- Purity:>90%
- Molecular Weight:46.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together