Human Recombinant TNF-alpha (from HEK293)
Marketsource
Supplier: ACRO Biosystems
About this item
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 103737-588
- 103015-544
- Protein/Peptide Name:
- TNF-alpha
- TNF-alpha
- Protein Synonyms:
- DIF|TNFα|cachexin|cachectin|TNF-alpha|TNFSF2|TNFA
- DIF|TNFα|cachexin|cachectin|TNF-alpha|TNFSF2|TNFA
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- P01375-1
- P01375-1
- Protease-free:
- Y
- Y
- Animal-Free:
- Y
- Y
- Nuclease-free:
- Y
- Y
- Carrier-Free:
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Endotoxin Content:
- Less than 0.1 EU per μg by the LAL method.
- Less than 0.1 EU per μg by the LAL method.
- Biological Activity:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Humira with a linear range of 0.2-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, Fc Tag (Cat. No. TN1-H5251) with a linear range of 0.2-3 ng/mL (Routinely tested).; Humira (Adalimumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) with an affinity constant of 0.255 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) induces cytotoxicity effect on the WEH1-13VAR cells in the presence of the metabolic inhibitor actinomycin D. The EC50 for this effect is 0.007-0.014 ng/mL (Routinely tested).; Neutralization assay shows that the cytotoxicity effect of ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) was inhibited by increasing concentration of Adalimumab. The concentration of TNF-alpha used is 1 ng/mL. The IC50 is 29 ng/mL (Routinely tested).
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Humira with a linear range of 0.2-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, Fc Tag (Cat. No. TN1-H5251) with a linear range of 0.2-3 ng/mL (Routinely tested).; Humira (Adalimumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) with an affinity constant of 0.255 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) induces cytotoxicity effect on the WEH1-13VAR cells in the presence of the metabolic inhibitor actinomycin D. The EC50 for this effect is 0.007-0.014 ng/mL (Routinely tested).; Neutralization assay shows that the cytotoxicity effect of ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) was inhibited by increasing concentration of Adalimumab. The concentration of TNF-alpha used is 1 ng/mL. The IC50 is 29 ng/mL (Routinely tested).
- Purity:
- >95%
- >95%
- Formulation:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Molecular Weight:
- 17.4 kDa
- 17.4 kDa
- Storage Conditions:
- –20 °C
- –20 °C
- Shipping Temperature:
- Powder,RT
- Powder,RT
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 25 µgDiscontinued-Supplier Number: TNAH421125UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103737-588Protein/Peptide NameTNF-alphaProtein SynonymsDIF|TNFα|cachexin|cachectin|TNF-alpha|TNFSF2|TNFAProtein/Peptide TypeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDP01375-1Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 0.1 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Humira with a linear range of 0.2-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, Fc Tag (Cat. No. TN1-H5251) with a linear range of 0.2-3 ng/mL (Routinely tested).; Humira (Adalimumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) with an affinity constant of 0.255 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) induces cytotoxicity effect on the WEH1-13VAR cells in the presence of the metabolic inhibitor actinomycin D. The EC50 for this effect is 0.007-0.014 ng/mL (Routinely tested).; Neutralization assay shows that the cytotoxicity effect of ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) was inhibited by increasing concentration of Adalimumab. The concentration of TNF-alpha used is 1 ng/mL. The IC50 is 29 ng/mL (Routinely tested).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight17.4 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource - Supplier Number: TNAH42111MGSpecifications:Marketsource
More
Specifications
Cat. No.103015-544Protein/Peptide NameTNF-alphaProtein SynonymsDIF|TNFα|cachexin|cachectin|TNF-alpha|TNFSF2|TNFAProtein/Peptide TypeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDP01375-1Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 0.1 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Humira with a linear range of 0.2-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (HPLC-verified) (Cat. No. TNA-H4211) at 2 μg/mL (100 μL/well) can bind Human TNFR1, Fc Tag (Cat. No. TN1-H5251) with a linear range of 0.2-3 ng/mL (Routinely tested).; Humira (Adalimumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) with an affinity constant of 0.255 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) induces cytotoxicity effect on the WEH1-13VAR cells in the presence of the metabolic inhibitor actinomycin D. The EC50 for this effect is 0.007-0.014 ng/mL (Routinely tested).; Neutralization assay shows that the cytotoxicity effect of ActiveMax® Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (Cat. No. TNA-H4211) was inhibited by increasing concentration of Adalimumab. The concentration of TNF-alpha used is 1 ng/mL. The IC50 is 29 ng/mL (Routinely tested).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight17.4 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



