Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Siglec-3 (from HEK293)
Human Recombinant Siglec-3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant Siglec-3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human CD33 MAb, Mouse IgG1 (Clone # 6C5/2) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Siglec-3, Avitag,His Tag (Cat. No. CD3-H82E7) with a linear range of 0.078-2.5 ng/mL (QC tested).
  • Gene ID:
    P20138
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    gp67|SIGLEC3|CD33
  • Protein/Peptide Name:
    Siglec-3
  • Purity:
    >90%
  • Molecular Weight:
    29.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together