Human Recombinant Siglec-2 (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human CD22 MAb at 0.5 μg/mL (100 μL/well) can bind Biotinylated Human Siglec-2, Fc,Avitag (Cat. No. SI2-H82F8) with a linear range of 1-16 ng/mL (QC tested).; 293 cells were transfected with anti-CD22-scFv and RFP tag. 2e5 of the cells were first stained with B. Biotinylated Human Siglec-2, Fc,Avitag (Cat. No. SI2-H82F8, 10 μg/mL) and C. Biotinylated Protein Control, followed by FITC Streptavidin. A. Non-transfected 293 cells and C. Biotinylated Protein Control were used as negative control. RFP was used to evaluate CAR (anti-CD22-scFv) expression and FITC was used to evaluate the binding activity of Biotinylated Human Siglec-2, Fc,Avitag (Cat. No. SI2-H82F8).; Human T cells were lentivirally transduced with anti-CD22 CAR and cultured for 3 days. Three days post-transduction, 1e6 cells were first incubated with 100 μL Biotinylated Human Siglec-2, Fc,Avitag (Cat. No. SI2-H82F8, 10 μg/mL), washed and then stained with PE Streptavidin. Non-transduced T cells were used as a control for gating of CAR expression. (Data are kindly provided by Cellyan Therapeutics Co. Ltd.)
- Gene ID:P20273-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:BL-CAM|SIGLEC-2|CD22|Siglec2|SIGLEC2|SIGLEC2FLJ22814
- Protein/Peptide Name:Siglec-2
- Purity:>95%
- Molecular Weight:103.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT