Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PD-1 (from HEK293)
Human Recombinant PD-1 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant PD-1 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) with a linear range of 0.02-0.32 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Nivolumab at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) with a linear range of 0.2-6 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) with a linear range of 20-156 ng/mL (Routinely tested).; Serial dilutions of nivolumab were added into Human PD-L1, Mouse IgG1 Fc Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H52A3): Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) binding reactions. The half maximal inhibitory concentration (IC50) is 0.3789 μg/mL (Routinely tested).; Serial dilutions of nivolumab were added into Human PD-L2, Mouse IgG1 Fc Tag (Cat. No. PD2-H52A5): Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) binding reactions. The half maximal inhibitory concentration (IC50) is 0.2823 μg/mL (Routinely tested).; Loaded Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) on SA Biosensor, can bind Human PD-L1, His Tag (HPLC verified) (Cat. No. PD1-H5229) with an affinity constant of 2.5 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human PD-1, Fc,Avitag (Cat. No. PD1-H82F1) on SA Biosensor, can bind Human PD-L2, His Tag (SPR verified) (Cat. No. PD2-H5220) with an affinity constant of 0.31 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q15116-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD279|PDCD1|PD1|SLEB2
  • Protein/Peptide Name:
    PD-1
  • Purity:
    >95%
  • Molecular Weight:
    44.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together