Human Recombinant PD-1 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) at 10 μg/mL (100 μL/well) can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) with a linear range of 2-40 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) at 2 μg/mL (100 μL/well) can bind Human PD-L2, Fc Tag (Cat. No. PD2-H5251) with a linear range of 0.2-4 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) at 2 μg/mL (100 μL/well) can bind Nivolumab with a linear range of 0.1-2 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, His Tag (HPLC-verified) (Cat. No. PD1-H5229) at series of concentration on MonRabTM Anti-His Tag (C-term) Antibody precoated (0.1 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) at 5 μg/mL (100 μL/well) with a linear range of 10-78 ng/mL (Routinely tested).; Loaded Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) on Protein A Biosensor, can bind Human PD-L1, His Tag (HPLC verified) (Cat. No. PD1-H5229) with an affinity constant of 2.6 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) on Protein A Biosensor, can bind Human PD-L2, His Tag (SPR verified) (Cat. No. PD2-H5220) with an affinity constant of 0.43 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q15116-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD279|PDCD1|PD1|SLEB2
- Protein/Peptide Name:PD-1
- Purity:>95%
- Molecular Weight:42.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together