Human Recombinant OX40 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.; Loaded Biotinylated Human OX40, Avitag,His Tag (Cat. No. TN4-H82E4) on SA Biosensor, can bind Anti-OX40 MAb (Human IgG1) with an affinity constant of 0.115 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human OX40, Avitag,His Tag (Cat. No. TN4-H82E4) on SA Biosensor, can bind Human OX40 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. OXL-H52Q8) with an affinity constant of 0.127 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P43489-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/Peptide Name:OX40
- Purity:>95%
- Molecular Weight:22.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together