Human Recombinant OX40 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human OX40 Ligand, His Tag (Cat. No. OXL-H52Q8) at 2 μg/mL (100 μL/well) can bind Human OX40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. OX0-H5252) with a linear range of 0.78-25 ng/mL (QC tested).; FACS assay shows that recombinant Human OX40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. OX0-H5252) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 is 0.01 μg/mL (Routinely tested).
- Gene ID:P43489-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/Peptide Name:OX40
- Purity:>95%
- Molecular Weight:47.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together