Human Recombinant Neuropilin-1 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human VEGF165, Tag Free (HPLC-verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Neuropilin-1, His,Avitag (Cat. No. NR1-H82E3) with a linear range of 0.5-16 ng/mL (QC tested).
- Gene ID:O14786-1 (V179A)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:NRP1|Neuropilin-1|VEGF165R|NRP|CD304
- Protein/Peptide Name:Neuropilin-1
- Purity:>95%
- Molecular Weight:73.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT