Human Recombinant Nectin-3 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Nectin-1, His Tag (Cat. No. PV1-H5223) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.078-0.625 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.102-4 μg/mL (Routinely tested).
- Gene ID:Q9NQS3-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD113|Nectin-3|PVRL3
- Protein/Peptide Name:Nectin-3
- Purity:>95%
- Molecular Weight:66.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together