Human Recombinant Nectin-2 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human PVRIG, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PVG-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.2-13 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.313-10 μg/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PVRIG, Fc Tag (Cat. No. PVG-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.2-16 ng/mL (Routinely tested).; Serial dilutions of TIGIT antibody antibody (1:2 serial dilutions, from 20 μg/mL to 0.0097 μg/mL) were added into Human TIGIT, Mouse IgG2a Fc Tag (Cat. No. TIT-H5253): Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) binding reactions. The assay was performed according to the above described protocol. Background was subtracted from data points before curve fitting.
- Gene ID:Q92692-2
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Nectin-2|PVRL2|CD112
- Protein/Peptide Name:Nectin-2
- Purity:>95%
- Molecular Weight:64.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together