Human Recombinant Mesothelin (from HEK293)
103738-012
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.01 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Mesothelin (296-580), His Tag, low endotoxin (Cat. No. MSN-H522a) at 0.5 μg/mL (100 μL/well) can bind Anti-MSLN MAb, Human IgG1 with a linear range of 0.2-3 ng/mL (QC tested).
- Gene ID:Q13421
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:MPF|MSLN|Mesothelin
- Protein/Peptide Name:Mesothelin
- Purity:>95%
- Molecular Weight:33.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-012