Human Recombinant Mesothelin (from HEK293)
Marketsource
Supplier: ACRO Biosystems
About this item
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 103738-272
- 103738-270
- Protein/Peptide Name:
- Mesothelin
- Mesothelin
- Protein Synonyms:
- MPF|MSLN|Mesothelin
- MPF|MSLN|Mesothelin
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- HEK293 cells
- HEK293 cells
- Conjugation:
- Unconjugated
- Unconjugated
- Gene ID:
- Q13421-3 (R309P)
- Q13421-3 (R309P)
- Protease-free:
- Y
- Y
- Animal-Free:
- Y
- Y
- Nuclease-free:
- Y
- Y
- Carrier-Free:
- Y
- Y
- Endotoxin Level:
- Low
- Low
- Endotoxin Content:
- Less than 0.1 EU per μg by the LAL method.
- Less than 0.1 EU per μg by the LAL method.
- Biological Activity:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253, 1 µg/ml) and C. Fc Tag Fusion Protein Control, followed by FITC anti-human IgG Fc. A. Non-transfected 293 cells and C. Fc Tag Fusion Protein Control were used as negative control. RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253, 1 µg/ml) and C. Fc Tag Fusion Protein Control, followed by FITC anti-human IgG Fc. A. Non-transfected 293 cells and C. Fc Tag Fusion Protein Control were used as negative control. RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).
- Purity:
- >95%
- >95%
- Formulation:
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Molecular Weight:
- 58.6 kDa
- 58.6 kDa
- Storage Conditions:
- –20 °C
- –20 °C
- Shipping Temperature:
- Powder,RT
- Powder,RT
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: MSNH5253100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103738-272Protein/Peptide NameMesothelinProtein SynonymsMPF|MSLN|MesothelinProtein/Peptide TypeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDQ13421-3 (R309P)Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 0.1 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253, 1 µg/ml) and C. Fc Tag Fusion Protein Control, followed by FITC anti-human IgG Fc. A. Non-transfected 293 cells and C. Fc Tag Fusion Protein Control were used as negative control. RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight58.6 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource - Supplier Number: MSNH52531MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103738-270Protein/Peptide NameMesothelinProtein SynonymsMPF|MSLN|MesothelinProtein/Peptide TypeRecombinantSpeciesHumanSourceHEK293 cellsConjugationUnconjugatedGene IDQ13421-3 (R309P)Protease-freeYAnimal-FreeYNuclease-freeYCarrier-FreeYEndotoxin LevelLowEndotoxin ContentLess than 0.1 EU per μg by the LAL method.Biological ActivityLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 293 cells were transfected with anti-MSLN-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253, 1 µg/ml) and C. Fc Tag Fusion Protein Control, followed by FITC anti-human IgG Fc. A. Non-transfected 293 cells and C. Fc Tag Fusion Protein Control were used as negative control. RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H5253).Purity>95%FormulationLyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.Molecular Weight58.6 kDaStorage Conditions–20 °CShipping TemperaturePowder,RTMarketsource



