Human Recombinant M-CSF (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human M-CSF R, Fc Tag (Cat. No. CSR-H5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) with a linear range of 0.1-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) at 2 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Cynomolgus M-CSF R, Fc Tag (Cat. No. CSR-C5252) with a linear range of 1-39 ng/mL (Routinely tested).; Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) immobilized on SA Chip can bind Human M-CSF R, His Tag (Cat. No. CSR-H5228) with an affinity constant of 0.309 μM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Biotinylated Human M-CSF, His,Avitag (Cat. No. MCF-H82E6) on SA Biosensor, can bind Human M-CSF R, Fc Tag, low endotoxin (Cat. No. CSR-H5258) with an affinity constant of 24.5 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P09603-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CSF-1|M-CSF|Lanimostim
- Protein/Peptide Name:M-CSF
- Purity:>90%
- Molecular Weight:28.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT