Human Recombinant LRRC32 (from HEK293)
Catalog # 103737-902
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Loaded Human LRRC32, Fc Tag (Cat. No. LR2-H5256) on AHC Biosensor, can bind Human TGFB1, His Tag (Cat. No. TG1-H524x) with an affinity constant of 0.288 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).
- Gene ID:Q14392-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:GARP|D11S833E|Garpin
- Protein/Peptide Name:LRRC32
- Purity:>95%
- Molecular Weight:92.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103737-902