Human Recombinant LAIR-2 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Biotinylated Human LAIR-2, His,Avitag (Cat. No. LA2-H82E4) with a linear range of 2-78 ng/mL (QC tested).
- Gene ID:Q6ISS4-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD306|LAIR2
- Protein/Peptide Name:LAIR-2
- Purity:>95%
- Molecular Weight:17.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together