Human Recombinant LAG-3 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human LAG3 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human LAG-3, Fc,Avitag (Cat. No. LA3-H82Fb) with a linear range of 0.1-3 ng/mL (QC tested).; Serial dilutions of Anti-LAG3, Neutralizing Antibody were added into Human FGL1, His Tag (Cat. No. FG1-H52Hy): Biotinylated Human LAG-3, Fc,Avitag (Cat. No. LA3-H82Fb) binding reactions. The half maximal inhibitory concentration (IC50) is 0.16695 μg/mL (Routinely tested).
- Gene ID:P18627-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FDC|CD223|LAG3
- Protein/Peptide Name:LAG-3
- Purity:>95%
- Molecular Weight:75.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together