Human Recombinant IL-7 (from HEK293)
Catalog # 103739-086
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) at 2 μg/mL (100 μL/well) can bind Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) with a linear range of 1-16 ng/mL (QC tested).; ActiveMax® Human IL-7, Tag Free; Captured Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) on CM5 chip via anti-human IgG Fc antibodies surface can bind ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) with an affinity constant of 23.9 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Captured Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) on CM5 chip via anti-mouse antibodies surface can bind ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) with an affinity constant of 13.3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) stimulates proliferation of PHA-P-activated human peripheral blood mononuclear cell (PBMC). The ED50 for this effect is 0.108-0.829 ng/mL (Routinely tested).
- Gene ID:P13232
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL7|Interleukin-7
- Protein/Peptide Name:IL-7
- Purity:>95%
- Molecular Weight:17.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-086