Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant ICAM-1 (from HEK293)
Human Recombinant ICAM-1 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant ICAM-1 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P05362-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    BB2|CD54|P3.58|ICAM1
  • Protein/Peptide Name:
    ICAM-1
  • Purity:
    >95%
  • Molecular Weight:
    51.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together