Human Recombinant HMGB1 (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Human HMGB1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. HM1-H5255) stimulates production of TNF-α in Raw-246.7 cells. The EC50 for this effect is 13.18 μg/mL (Routinely tested).
- Gene ID:P09429-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:SBP-1|HMGB1|HMG3|HMG1
- Protein/Peptide Name:HMGB1
- Purity:>95%
- Molecular Weight:51.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together