Human Recombinant Gremlin (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5 with 1 mM EDTA. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human BMP-2, Tag Free (Cat. No. BM2-H4117) at 2μg/mL (100 μL/well) can bind Human Gremlin, Fc Tag (Cat. No. GR1-H5254) with a linear range of 0.8-80 ng/mL (QC tested).
- Gene ID:O60565
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CKTSF1B1|DRM|DAND2|PIG2|GREM1
- Protein/Peptide Name:Gremlin
- Purity:>95%
- Molecular Weight:44.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5 with 1 mM EDTA. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together