Human Recombinant Glypican 3 (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human GPC3 MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) with a linear range of 0.078-1.25 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).; 293 cells were transfected with anti-GPC3-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Glypican 3 Protein, Fc Tag (Cat. No. GP3-HF258, 3 µg/ml) and C. FITC-labeled Protein Control. A. Non-transfected 293 cells and C. FITC-labeled Protein Control were used as negative control. RFP was used to evaluate CAR (anti-GPC3-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human Glypican 3 Protein, Fc Tag (Cat. No. GP3-HF258).
- Gene ID:P51654-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
- Protein/Peptide Name:Glypican 3
- Purity:>90%
- Molecular Weight:38.1 kDa and 49.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT, Avoid light