Human Recombinant Erythropoietin (from HEK293)
Catalog # 103739-032
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P01588
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:EP|Hemopoietin|EPO|Erthropoyetin|Erythropoietin|Erythropoetin|Hematopoietin|MVCD2
- Protein/Peptide Name:Erythropoietin
- Purity:>97%
- Molecular Weight:18.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-032