Human Recombinant CTLA-4 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.1-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.5-7.8 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Ipilimumab at 5 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.1-0.8 ng/mL (Routinely tested).; Loaded Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) on Protein A Biosensor, can bind Human B7-1, His Tag (Cat. No. B71-H5228) with an affinity constant of 0.52 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) on Protein A Biosensor, can bind Human B7-2, His Tag (Cat. No. CD6-H5223) with an affinity constant of 1.6 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P16410-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CTLA4|CD152
- Protein/Peptide Name:CTLA-4
- Purity:>95%
- Molecular Weight:40.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together