Human Recombinant CEACAM-8 (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CEACAM-6, His Tag (Cat. No. CE6-H5223) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-8, His,Avitag (Cat. No. CE8-H82E9) with a linear range of 0.02-0.313 μg/mL (QC tested).
- Gene ID:P31997-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CEACAM8|CD66b|NCA-95|CGM6|CD67
- Protein/Peptide Name:CEACAM-8
- Purity:>95%
- Molecular Weight:35.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT