Human Recombinant CD47 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.; Loaded Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) on Protein A Biosensor, can bind Human SIRP alpha, His Tag (HPLC-verified) (Cat. No. SIA-H5225) with an affinity constant of 1.1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; FACS assay shows that Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) can bind to ACHN cell expressing human SIRP-a. The concentration of CD47 used is 10 μg/mL (Routinely tested).; FACS analysis shows that the binding of Human CD47 to ACHN expressing SIRP-a was inhibited by increasing concentration of neutralizing SIRP-a antibody. The concentration of Human CD47 used is 10 μg/mL. IC50=8.39 μg/mL
- Gene ID:Q08722
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IAP|MER6|OA3
- Protein/Peptide Name:CD47
- Purity:>95%
- Molecular Weight:40.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together